missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyl-N-[1-(2-phenylethyl)piperidin-4-yl]acetamide, Mikromol

Product Code. 30463936
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463936 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463936 Supplier Mikromol Supplier No. MM0528.03

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Phenyl-N-[1-(2-phenylethyl)piperidin-4-yl]acetamide, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetyl Fentanyl
CAS 3258-84-2
Analyte or Component Names Acetyl Fentanyl
Shipping Condition Room Temperature
Molecular Formula C21 H26 N2 O
Certifications/Compliance ISO 17025
Synonym Fentanyl Imp. C (EP), Fentanyl USP Related Compound G CI, Fentanyl USP RC G CI, N-Phenyl-N-[1-(2-phenylethyl)piperidin-4-yl]acetamide, N-Phenyl-N-[1-(2-phenylethyl)-4-piperidinyl]acetamide, N-(1-Phenethyl-4-piperidyl)acetanilide, 1-Phenethyl-4-(N-phenylacetamido)piperidine, Acetylfentanyl, Fentanyl Related Compound G, Fentanyl Citrate Impurity C, Fentanyl Impurity C, Fentanyl Impurity C
InChI Formula InChI=1S/C21H26N2O/c1-18(24)23(20-10-6-3-7-11-20)21-13-16-22(17-14-21)15-12-19-8-4-2-5-9-19/h2-11,21H,12-17H2,1H3
Solution Type Neat
SMILES CC(=O)N(C1CCN(CCc2ccccc2)CC1)c3ccccc3
IUPAC Name N-phenyl-N-[1-(2-phenylethyl)piperidin-4-yl]acetamide
Molecular Weight (g/mol) 322.44
Quantity 100 mg
Formula Weight 322.2045 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.