missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylacetamide (Acetanilide), LoGiCal

Product Code. 30459273
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459273 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459273 Supplier LoGiCal Supplier No. LGCFOR0042.10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Phenylacetamide
CAS 103-84-4
Analyte or Component Names N-Phenylacetamide
Shipping Condition Room Temperature
Molecular Formula C8 H9 N O
Certifications/Compliance ISO 9001
Synonym N-Phenylacetamide, Paracetamol Imp. D (EP), Acetanilide, Acetamidobenzene, Acetanil, Acetoanilide, Acetylaniline, Antifebrin, Benzenamine, N-acetyl-, N-Acetylaminobenzene, N-Acetylaniline, NSC 203231, NSC 7636, Phenalgene, Phenalgin, Acetaminophen USP Related Compound D, Acetaminophen USP RC D, Acetaminophen Related Compound D, Paracetamol Impurity D
InChI Formula InChI=1S/C8H9NO/c1-7(10)9-8-5-3-2-4-6-8/h2-6H,1H3,(H,9,10)
Solution Type Neat
SMILES CC(=O)Nc1ccccc1
IUPAC Name N-phenylacetamide
Industry Type Clinical Testing
Molecular Weight (g/mol) 135.16
Quantity 10 mg
Formula Weight 135.0684 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.