missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylpropanamide, Mikromol™

Product Code. 17960466
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17960466 25 mg 25mg
17901646 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17960466 Supplier Mikromol Supplier No. MM0528.070025
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Phenylpropanamide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Phenylpropanamide
CAS 620-71-3
Analyte or Component Names N-Phenylpropanamide
Shipping Condition Room Temperature
Molecular Formula C9 H11 N O
Certifications/Compliance ISO 17025
Synonym Fentanyl Imp. G (EP), N-Phenylpropanamide, Fentanyl Impurity G, Remifentanil Hydrochloride Impurity O, Remifentanil Impurity O, Remifentanil Impurity O
InChI Formula InChI=1S/C9H11NO/c1-2-9(11)10-8-6-4-3-5-7-8/h3-7H,2H2,1H3,(H,10,11)
SMILES CCC(=O)Nc1ccccc1
IUPAC Name N-phenylpropanamide
Molecular Weight (g/mol) 149.19
Quantity 25 mg
Formula Weight 149.0841 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.