missing translation for 'onlineSavingsMsg'
Learn More

N-[(R)-1-Carbethoxybutyl]-(S)-alanine, TRC

Product Code. 30361148
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361148 250 mg 250mg
30355159 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30361148 Supplier Toronto Research Chemicals Supplier No. TRCC178000250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(R)-1-Carbethoxybutyl]-(S)-alanine
Analyte or Component Names N-[(R)-1-Carbethoxybutyl]-(S)-alanine
Shipping Condition Room Temperature
Molecular Formula C10H19NO4
Synonym (S)-2-(((R)-1-Ethoxy-1-oxopentan-2-yl)amino)propanoic acid
InChI Formula InChI=1S/C10H19NO4/c1-4-6-8(10(14)15-5-2)11-7(3)9(12)13/h7-8,11H,4-6H2,1-3H3,(H,12,13)/t7-,8+/m0/s1
Solution Type Neat
SMILES O=C(O)[C@H](C)N[C@@H](C(OCC)=O)CCC
IUPAC Name (2S)-2-[[(2R)-1-ethoxy-1-oxopentan-2-yl]amino]propanoic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 217.26
Quantity 250 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.