missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N4BP1 Antibody (SR1060), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22555-50ul
This item is not returnable.
View return policy
Description
N4BP1 Monoclonal antibody specifically detects N4BP1 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| N4BP1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot | |
| SR1060 | |
| Western Blot 1:500-1:2000 | |
| FLJ31821, KIAA0615, MGC176730, NEDD4 binding protein 1, NEDD4-binding protein 1 | |
| A synthesized peptide derived from human N4BP1 (Uniprot #: Q6A037) | |
| 50 μg | |
| Cancer, Cell Cycle and Replication | |
| 9683 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction