missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N4BP1 Antibody (SR1060), Novus Biologicals™
Rabbit Monoclonal Antibody
338.00 € - 449.00 €
Specifications
| Antigen | N4BP1 |
|---|---|
| Clone | SR1060 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18645204
|
Novus Biologicals
NBP3-22555-100ul |
100 μg |
449.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18621265
|
Novus Biologicals
NBP3-22555-50ul |
50 μg |
338.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N4BP1 Monoclonal antibody specifically detects N4BP1 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| N4BP1 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| FLJ31821, KIAA0615, MGC176730, NEDD4 binding protein 1, NEDD4-binding protein 1 | |
| A synthesized peptide derived from human N4BP1 (Uniprot #: Q6A037) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1060 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 9683 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title