missing translation for 'onlineSavingsMsg'
Learn More

N6-methyladenosine (m6A) Antibody (7N6C7) [DyLight 488], Novus Biologicals Biologicals™

Product Code. 30494739 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30494739 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30494739 Supplier Novus Biologicals Supplier No. NBP333194G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

N6-methyladenosine (m6A) Monoclonal antibody specifically detects N6-methyladenosine (m6A) in Non-species specific samples. It is validated for ELISA,Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen N6-methyladenosine (m6A)
Applications ELISA, Dot Blot
Classification Monoclonal
Clone 7N6C7
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias (-)-6-(Methylamino)purine riboside, N(6)-Methyladenosine, N-Methyladenosine
Host Species Mouse
Immunogen Chemical compounds corresponding to N6-methyladenosine (m6A).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.