missing translation for 'onlineSavingsMsg'
Learn More

Naltrexone Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30461590
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461590 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461590 Supplier LoGiCal Supplier No. LGCAMP0233.0001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Naltrexone Hydrochloride 1.0 mg/ml in Methanol (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Naltrexone hydrochloride
CAS 16676-29-2
Analyte or Component Names Naltrexone hydrochloride
Shipping Condition Ice Pack (-20°C)
Molecular Formula C20 H23 N O4 . Cl H
Certifications/Compliance ISO 17034
Synonym Antaxone, Depade, EN 1639 A, NIH 8503, Nalorex, Morphinan-6-one, 17-(cyclopropylmethyl)-4,5 alpha-epoxy-3,14-dihydroxy-, hydrochloride (8 CI), Morphinan-6-one, 17-(cyclopropylmethyl)-4,5 alpha-epoxy-3,14-dihydroxy-, hydrochloride (8 CI), Morphinan-6-one, 17-(cyclopropylmethyl)-4,5 alpha-epoxy-3,14-dihydroxy-, hydrochloride (8 CI), Morphinan-6-one, 17-(cyclopropylmethyl)-4,5 alpha-epoxy-3,14-dihydroxy-, hydrochloride (8 CI), Morphinan-6-one, 17-(cyclopropylmethyl)-4,5 alpha-epoxy-3,14-dihydroxy-, hydrochloride (8 CI), Morphinan-6-one, 17-(cyclopropylmethyl)-4,5 alpha-epoxy-3,14-dihydroxy-, hydrochloride (8 CI)
InChI Formula InChI=1 S/C20H23NO4.ClH/c22-13-4-3-12-9-15-20(24)6-5-14(23)18-19(20,16(12)17(13)25-18)7-8-21(15)10-11-1-2-11;/h3-4,11,15,18,22,24 H,1-2,5-10H2;1 H/t15-,18+,19+,20-;/m1./s1
SMILES Cl.Oc1ccc2C[C@H]3 N(CC4CC4)CC[C@@]56 [C@@H](Oc1c25)C(=O)CC[C@@]36 O
IUPAC Name (4 R,4 AS,7 aR,12 bS)-3-(cyclopropylmethyl)-4 A,9-dihydroxy-2,4,5,6,7 A,13-hexahydro-1 H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;hydrochloride
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 377.86
Quantity 1 mL
Formula Weight 377.1394 g/mol
Show More Show Less
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.