missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naphazoline Nitrate, Mikromol
Description
Naphazoline Nitrate, Mikromol
Specifications
Specifications
| Chemical Name or Material | Naphazoline nitrate |
| CAS | 5144-52-5 |
| Analyte or Component Names | Naphazoline nitrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H14 N2 . H N O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | 1H-Imidazole, 4,5-dihydro-2-(1-naphthalenylmethyl)-, nitrate (1:1), 1H-Imidazole, 4,5-dihydro-2-(1-naphthalenylmethyl)-, mononitrate (9CI), 2-Imidazoline, 2-(1-naphthylmethyl)-, mononitrate (8CI), Naftizin, Naphazoline nitrate, Naphthisin, Naphthizin, Naphthylmethyl-2-imidazoline nitrate, Naphtyzin, Privina, Privine, Privine (British), Privine nitrate, Rinazin, Sanorin |
| InChI Formula | InChI=1S/C14H14N2.HNO3/c1-2-7-13-11(4-1)5-3-6-12(13)10-14-15-8-9-16-14;2-1(3)4/h1-7H,8-10H2,(H,15,16);(H,2,3,4) |
| Solution Type | Neat |
| SMILES | O[N+](=O)[O-].C(C1=NCCN1)c2cccc3ccccc23 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?