missing translation for 'onlineSavingsMsg'
Learn More

NAT10 Antibody [Allophycocyanin], Novus Biologicals Biologicals™

Product Code. 30512085 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30512085 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30512085 Supplier Novus Biologicals Supplier No. NBP338407APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NAT10 Polyclonal antibody specifically detects NAT10 in Human,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAT10
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias ALP, DKFZp434C116, EC 2.3.1.-, FLJ10774, FLJ12179, FLJ23850, hALP, KIAA1709, N-acetyltransferase 10, N-acetyltransferase 10 (GCN5-related), N-acetyltransferase-like protein, NET43
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-250 of human NAT10 (NP_078938.2).,, Sequence:, DAPAHHLFCLLPPVPPTQNALPEVLAVIQVCLEGEISRQSILNSLSRGKKASGDLIPWTVSEQFQDPDFGGLSGGRVVRIAVHPDYQGMGYGSRALQLLQMYYEGRFPCLEEKVLETPQEIHTVSSEAVSLLEEVITPRKDLPPLLLKLNERPAERLDYLGVSYGLTPRLLKFWKRAGFVPVYLRQTPNDLTGEHSCIMLKTLT
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Direct Reversal of DNA Damage, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 55226
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.