missing translation for 'onlineSavingsMsg'
Learn More

Nav1.6 Rabbit anti-Human, Polyclonal, R&D Systems™

Product Code. 18366502 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18366502 25 μg 25µL
18321326 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18366502 Supplier Novus Biologicals Supplier No. NBP31727925UL
  • ONLINE EXCLUSIVE PRICE
    538.16 €
    458.85 € / 25µL
    Save 79.31 €
    14% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

Nav1.6 Polyclonal specifically detects Nav1.6 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nav1.6
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias CerIII, hNa6/Scn8a voltage-gated sodium channel, MED, NaCh6, Nav1.6, PN4, sodium channel protein type 8 subunit alpha, Sodium channel protein type VIII subunit alpha, sodium channel, voltage gated, type VIII, alpha polypeptide, sodium channel, voltage gated, type VIII, alpha subunit, Voltage-gated sodium channel subunit alpha Nav1.6
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the amino acids: QEEVSAVVLQRAYRGHLARRGFICKKTTSNKLENGGTHREKKESTPSTASLPSYDSV
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6334
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.