missing translation for 'onlineSavingsMsg'
Learn More

NCALD Antibody - BSA Free, Novus Biologicals™

Product Code. 18643131 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.01mL
0.02mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18643131 0.02 mL 0.02mL
18640681 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18643131 Supplier Novus Biologicals Supplier No. NBP2936930.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NCALD Polyclonal antibody specifically detects NCALD in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCALD
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias MGC33870, MGC74858, neurocalcin delta, neurocalcin-delta
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-193 of human NCALD (NP_114430.2). MGKQNSKLRPEVMQDLLESTDFTEHEIQEWYKGFLRDCPSGHLSMEEFKKIYGNFFPYGDASKFAEHVFRTFDANGDGTIDFREFIIALSVTSRGKLEQKLKWAFSMYDLDGNGYISKAEMLEIVQAIYKMVSSVMKMPEDESTPEKRTEKIFRQMDTNRDGKLSLEEFIRGAKSDPSIVRLLQCDPSSAGQF
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 83988
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.