missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDRG1 Mouse, Clone: NDRG1/1383, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 10397-MSM1-P1
This item is not returnable.
View return policy
Specifications
| NDRG1 | |
| Cytoplasmic and Cell Surface | |
| NDRG1/1383 | |
| Unconjugated | |
| XP_011515093 | |
| Mouse | |
| 43kDa | |
| Protein A/G Purified | |
| RUO | |
| 10397 | |
| IgG3 |
| Western Blot | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| NDRG1 | |
| Recombinant fragment of human NDRG1 fragment (aa162-283) | |
| HeLa Cells. Prostrate or Renal Carcinoma. | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction