missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDRG1 Mouse, Clone: NDRG1/1383, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 478.00 €
Specifications
| Antigen | NDRG1 |
|---|---|
| Clone | NDRG1/1383 |
| Concentration | 200μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15929727
|
EnquireBio
10397-MSM1-P0 |
20 μg | |||||||
|
15939727
|
EnquireBio
10397-MSM1-P1 |
100 μg |
554.40 € 478.00 € / 100µg Save 76.40 € 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| NDRG1 | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| 10397 | |
| Recombinant fragment of human NDRG1 fragment (aa162-283) | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 43kDa |
| NDRG1/1383 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| XP_011515093 | |
| NDRG1 | |
| IgG3 | |
| Protein A/G Purified | |
| Cytoplasmic and Cell Surface | |
| HeLa Cells. Prostrate or Renal Carcinoma. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title