missing translation for 'onlineSavingsMsg'
Learn More

NDRG2 Antibody (4H3) [DyLight 594], Novus Biologicals Biologicals™

Product Code. 30519889 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30519889 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30519889 Supplier Novus Biologicals Supplier No. NBP343066DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NDRG2 Monoclonal antibody specifically detects NDRG2 in Human,Mouse,Porcine,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDRG2
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4H3
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias DKFZp781G1938, FLJ25522, KIAA1248cytoplasmic protein Ndr1, NDR1-related protein NDR2, NDRG family member 2, protein NDRG2, Protein Syld709613, syld709613 protein, SYLDN-myc downstream regulator 2
Host Species Mouse
Immunogen NDRG family member 2 - (Uniprot# Q9UN36)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 57447
Target Species Human, Mouse, Pig, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Show More Show Less
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.