missing translation for 'onlineSavingsMsg'
Learn More

NDUFA4L2 Antibody (0B11) [PE], Novus Biologicals Biologicals™

Product Code. 30521139 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30521139 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30521139 Supplier Novus Biologicals Supplier No. NBP343005PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NDUFA4L2 Monoclonal antibody specifically detects NDUFA4L2 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFA4L2
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 0B11
Conjugate PE
Formulation PBS
Gene Alias NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4-like 2, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 4-like 2, NADH:ubiquinone oxidoreductase MLRQ subunit homolog, NADH-ubiquinone oxidoreductase MLRQ subunit homolog, NUOMSFLJ26118
Host Species Mouse
Immunogen NADH dehydrogenase(ubiquinone) 1 alpha subcomplex, 4-like 2 - (Uniprot# Q9NRX3)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Endocrinology, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 56901
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.