missing translation for 'onlineSavingsMsg'
Learn More

NDUFB6 Antibody, Novus Biologicals™

Product Code. 18642908 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18642908 100 μg 100µL
18698498 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18642908 Supplier Novus Biologicals Supplier No. NBP312619100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NDUFB6 Polyclonal antibody specifically detects NDUFB6 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFB6
Applications Western Blot, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunoprecipitation 1-5ul/mg of lysate
Formulation PBS, pH7.0
Gene Alias B17, CI, CI-B17, complex I, mitochondrial respiratory chain, B17 subunit, Complex I-B17, MGC13675, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 6 (17kD, B17), NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 6, 17kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 6, NADH-ubiquinone oxidoreductase B17 subunit, NADH-ubiquinone oxidoreductase beta subunit, 6
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NDUFB6 fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4712
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.