missing translation for 'onlineSavingsMsg'
Learn More

Ndufs1 Antibody [mFluor Violet 610 SE], Novus Biologicals Biologicals™

Product Code. 30487105 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30487105 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30487105 Supplier Novus Biologicals Supplier No. NBP335706MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Ndufs1 Polyclonal antibody specifically detects Ndufs1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ndufs1
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias CI-75k, CI-75Kd, complex I 75kDa subunit, complex I, mitochondrial respiratory chain, 75-kD subunit, Complex I-75kD, EC 1.6.5.3, EC 1.6.99.3, MGC26839, mitochondrial NADH-ubiquinone oxidoreductase 75 kDa subunit, NADH dehydrogenase (ubiquinone) Fe-S protein 1 (75kD) (NADH-coenzyme Qreductase), NADH dehydrogenase (ubiquinone) Fe-S protein 1, 75kDa (NADH-coenzyme Qreductase), NADH-ubiquinone oxidoreductase 75 kDa subunit, mitochondrial, PRO1304
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 80-290 of human Ndufs1 (NP_004997.4).,, Sequence:, VEIEKAPKVVAACAMPVMKGWNILTNSEKSKKAREGVMEFLLANHPLDCPICDQGGECDLQDQSMMFGNDRSRFLEGKRAVEDKNIGPLVKTIMTRCIQCTRCIRFASEIAGVDDLGTTGRGNDMQVGTYIEKMFMSELSGNIIDICPVGALTSKPYAFTARPWETRKTESIDVMDAVGSNIVVSTRTGEVMRILPRMHEDINEEWISDKT
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 4719
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.