missing translation for 'onlineSavingsMsg'
Learn More

NDUFV2 Antibody [DyLight 650], Novus Biologicals Biologicals™

Product Code. 30512687 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30512687 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30512687 Supplier Novus Biologicals Supplier No. NBP338428C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NDUFV2 Polyclonal antibody specifically detects NDUFV2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFV2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias CI-24k, mitochondrial respitory chain, 24 kD subunit, NADH dehydrogenase (ubiquinone) flavoprotein 2 (24kD), NADH dehydrogenase (ubiquinone) flavoprotein 2, 24kDa, NADH dehydrogenase [ubiquinone] flavoprotein 2, mitochondrial, NADH dehydrogenase ubiquinone flavoprotein 2, mitochondrial, NADH-ubiquinone oxidoreductase 24 kDa subunit, NADH-ubiquinone oxidoreductase flavoprotein 2, nuclear-encoded mitochondrial NADH-ubiquinone reductase 24Kd subunit
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-249 of human NDUFV2 (NP_066552.2).,, Sequence:, MFFSAALRARAAGLTAHWGRHVRNLHKTVMQNGAGGALFVHRDTPENNPDTPFDFTPENYKRIEAIVKNYPEGHKAAAVLPVLDLAQRQNGWLPISAMNKVAEVLQVPPMRVYEVATFYTMYNRKPVGKYHIQVCTTTPCMLRNSDSILEAIQKKLGIKVGETTPDKLFTLIEVECLGACVNAPMVQINDNYYEDLTAKDIEEIIDELKAGKIPKPGPRSGRFSCEPAGGLTSLTEPPKGPGFGVQAGL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4729
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.