missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neomycin B-LP Acetate Salt Hydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Neomycin B-LP Acetate Salt Hydrate |
| Analyte or Component Names | Neomycin B-LP Acetate Salt Hydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25H48N6O14 . x(C 2H4O2) y(H2O) |
| InChI Formula | InChI=1S/C25H48N6O14.C2H4O2.H2O/c1-6(33)31-8-2-7(28)14(34)22(20(8)43-23-12(29)17(37)15(35)9(3-26)40-23)45-25-19(39)21(11(5-32)42-25)44-24-13(30)18(38)16(36)10(4-27)41-24;1-2(3)4;/h7-25,32,34-39H,2-5,26-30H2,1H3,(H,31,33);1H3,(H,3,4);1H2/t7-,8+,9-,10+,11-,12-,13-,14+,15-,16-,17-,18-,19-,20-,21-,22-,23-,24?,25+;;/m1../s1 |
| Solution Type | Neat |
| SMILES | O[C@H]1[C@H](O)[C@@H](N)[C@@H](O[C@H]2[C@H](O[C@@H]3O[C@H](CO)[C@@H](OC4[C@@H]([C@@H](O)[C@H](O)[C@H](CN)O4)N)[C@H]3O)[C@@H](O)[C@H](N)C[C@@H]2NC(C)=O)O[C@@H]1CN.CC(O)=O.O |
| IUPAC Name | N-((1S,2R,3R,4S,5R)-5-amino-2-(((2R,3R,4R,5S,6R)-3-amino-6-(aminomethyl)-4,5-dihydroxytetrahydro-2H-pyran-2-yl)oxy)-3-(((2S,3R,4S,5R)-4-(((3R,4R,5S,6S)-3-amino-6-(aminomethyl)-4,5-dihydroxytetrahydro-2H-pyran-2-yl)oxy)-3-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-4-hydroxycyclohexyl)acetamide acetate hydrate |
| Industry Type | Forensics |
| Molecular Weight (g/mol) | 734.7 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?