missing translation for 'onlineSavingsMsg'
Learn More

Nestin Antibody (10C2) - BSA Free, Novus Biologicals™

Product Code. 18737513 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18737513 0.025 mL 25µL
18771564 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18737513 Supplier Novus Biologicals Supplier No. NB300266SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 21 publication

Nestin Monoclonal specifically detects Nestin in Human, Mouse, Rabbit (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nestin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 10C2
Concentration 2.4 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:5000, Flow Cytometry 1 ug per million cells, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:100 - 1:200, Immunohistochemistry-Frozen 1:100 - 1:200, CyTOF-ready
Gene Accession No. P48681
Gene Alias FLJ21841, nestin
Gene Symbols NES
Host Species Mouse
Immunogen A fragment of the human Nestin protein corresponding to residues 1464-1614. [UniProt P48681]
Purification Method Protein G purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cancer Stem Cells, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 10763
Test Specificity Human. It does not cross-react mouse, rat or rabbit.
Target Species Human, Mouse, Rabbit (Negative), Rat (Negative)
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.