missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neurexin 3/NRXN3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 470.00 €
Specifications
| Antigen | Neurexin 3/NRXN3 |
|---|---|
| Dilution | Western Blot 1:1000-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18677300
|
Novus Biologicals
NBP2-93379-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18685490
|
Novus Biologicals
NBP2-93379-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neurexin 3/NRXN3 Polyclonal antibody specifically detects Neurexin 3/NRXN3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Neurexin 3/NRXN3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 9369 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| C14orf60, chromosome 14 open reading frame 60, KIAA0743MGC176711, neurexin 3, neurexin III, Neurexin III-alpha, Neurexin III-beta, neurexin-3-beta | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 373-432 of human NRXN3 (NP_620426.2). ILLYAMYKYRNRDEGSYQVDETRNYISNSAQSNGTLMKEKQQSSKSGHKKQKNKDREYYV | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title