missing translation for 'onlineSavingsMsg'
Learn More

Neurofibromin 1 Antibody (CL11773) [PE], Novus Biologicals Biologicals™

Product Code. 30531534 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30531534 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30531534 Supplier Novus Biologicals Supplier No. NBP344294PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Neurofibromin 1 Monoclonal antibody specifically detects Neurofibromin 1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurofibromin 1
Applications Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL11773
Conjugate PE
Formulation PBS
Gene Alias DKFZp686J1293, FLJ21220, Neurofibromatosis-related protein NF-1, neurofibromin, neurofibromin 1, NFNS, VRNF, WSS
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P21359, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Hypoxia, Neuroscience, Neurotransmission, Phospho Specific, Signal Transduction, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4763
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.