missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuronal Pentraxin 2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76899-0.025mg
This item is not returnable.
View return policy
Description
Neuronal Pentraxin 2 Polyclonal specifically detects Neuronal Pentraxin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| Neuronal Pentraxin 2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| NARP, neuronal activity-regulated pentaxin, neuronal pentraxin IIapexin, neuronal pentraxin-2, NP2, NP-II | |
| Rabbit | |
| 47 kDa | |
| 0.025 mg | |
| Stem Cell Markers | |
| 4885 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.5-4 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin | |
| NP_002514 | |
| NPTX2 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the center of the human NPTX2. The immunogen is located within amino acids 170 - 220 of NPTX2. Amino Acid Squence: LHNETSAHRQKTESTL | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction