missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuronal Pentraxin 2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
189.00 €
Specifications
| Antigen | Neuronal Pentraxin 2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-4 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Neuronal Pentraxin 2 Polyclonal specifically detects Neuronal Pentraxin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| Neuronal Pentraxin 2 | |
| Western Blot 0.5-4 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Rabbit | |
| Stem Cell Markers | |
| PBS with 0.02% Sodium Azide | |
| NARP, neuronal activity-regulated pentaxin, neuronal pentraxin IIapexin, neuronal pentraxin-2, NP2, NP-II | |
| NPTX2 | |
| IgG | |
| Affinity Purified | |
| 47 kDa |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| NP_002514 | |
| 4885 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the center of the human NPTX2. The immunogen is located within amino acids 170 - 220 of NPTX2. Amino Acid Squence: LHNETSAHRQKTESTL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title