missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neutrophils Antibody (RPN3/57), Novus Biologicals™
Mouse Monoclonal Antibody
188.00 € - 463.00 €
Specifications
| Antigen | Neutrophils |
|---|---|
| Clone | RPN3/57 |
| Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18684916
|
Novus Biologicals
NBP3-11629-0.025mg |
0.025 mg |
188.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627698
|
Novus Biologicals
NBP3-11629-0.1mg |
0.1 mg |
463.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Neutrophils Monoclonal antibody specifically detects Neutrophils in Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)Specifications
| Neutrophils | |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Rabbit | |
| Rabbit Peritoneal Neutrophils | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| RPN3/57 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title