Learn More
Thermo Scientific New Equipment Startup Service for Cold Storage, Freezer, Refrigerator, ULT AND Arctiko CRYO -150°C Compressor-based Freezer
Product Code. 32176308 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
ULT Freezers
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
32176308 ULT Freezers Each
1 options
Product Code. 32176308 Supplier Thermo Scientific Supplier No. STARTULTMVS
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Installation service scope covers compressor-based refrigerators, freezers, ULT and Arctiko CRYO -150°C equipment where LN2 backup is secondary to the primary cooling system.
  • Not suitable for liquid nitrogen Dewar-type cryopreservation systems
  • White-glove delivery, building works and primary LN2 cryopreservation systems are excluded unless separately quoted
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) ULT Freezers
Product Type Service