missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific New Equipment Startup Service for Recirculating Chiller

Product Code. 32176305 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Recirculating Chiller
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
32176305 Recirculating Chiller Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32176305 Supplier Thermo Scientific Supplier No. STARTCHILLMVS
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Installation service includes start-up assistance, basic commissioning checks and operator handover for recirculating chiller by trained Unity Lab Services engineers.

  • Standard local labour and travel within the agreed EMEA coverage area are included unless otherwise stated
  • Exceptional travel, non-standard site access, repeat visits caused by site readiness issues, LN2 backup-specific checks where relevant, and any work outside the agreed startup scope may be quoted separately
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Recirculating Chiller
Duration 1 Day
Product Type Service
Target Audience Employees/Users
Species All
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.