missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF-L Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP1-05219-0.025ML
This item is not returnable.
View return policy
Description
NF-L Polyclonal specifically detects NF-L in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.
Specifications
| NF-L | |
| Polyclonal | |
| Western Blot 1:5000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry Free-Floating 1:2000 | |
| P02548 | |
| NEFL | |
| Recombinant human NF-L protein | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Mouse, Human, Bovine, Rat, Pig | |
| Precipitation |
| Immunohistochemistry, Immunohistochemistry (Free Floating), Immunofluorescence, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| PBS with 5mM Sodium Azide | |
| 68 kDa neurofilament protein, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68FLJ53642, NF-L, NFLlight polypeptide 68kDa | |
| Chicken | |
| 68 kDa | |
| 0.025 mL | |
| Cellular Markers, Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience | |
| 4747 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction