missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NF-L Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
188.00 € - 499.00 €
Specifications
| Antigen | NF-L |
|---|---|
| Dilution | Western Blot 1:5000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry Free-Floating 1:2000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Free Floating), Immunofluorescence, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18232175
|
Novus Biologicals
NBP1-05219-0.025ML |
0.025 mL |
188.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18749043
|
Novus Biologicals
NBP1-05219 |
0.1 mL |
499.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NF-L Polyclonal specifically detects NF-L in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry Free-Floating.Specifications
| NF-L | |
| Immunohistochemistry, Immunohistochemistry (Free Floating), Immunofluorescence, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Chicken | |
| Cellular Markers, Cytoskeleton Markers, Neurodegeneration, Neurofilaments, Neuronal Cell Markers, Neuroscience | |
| P02548 | |
| 4747 | |
| Recombinant human NF-L protein | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:5000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry Free-Floating 1:2000 | |
| Polyclonal | |
| Precipitation | |
| RUO | |
| Mouse, Human, Bovine, Rat, Pig | |
| 68 kDa neurofilament protein, neurofilament protein, light chain, neurofilament subunit NF-L, Neurofilament triplet L protein, neurofilament, light polypeptide, neurofilament-light, NF68FLJ53642, NF-L, NFLlight polypeptide 68kDa | |
| NEFL | |
| IgY | |
| Ammonium sulfate precipitation | |
| 68 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title