missing translation for 'onlineSavingsMsg'
Learn More

NF-M Antibody (CL2697) [Alexa Fluor™ 700], Novus Biologicals™

Product Code. 30524083 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30524083 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30524083 Supplier Novus Biologicals Supplier No. NBP343821AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NF-M Monoclonal antibody specifically detects NF-M in Human,Mouse,Rat samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen NF-M
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL2697
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias NEF3neurofilament medium polypeptide, Neurofilament 3, neurofilament, medium polypeptide, neurofilament, medium polypeptide 150kDa, neurofilament-3 (150 kD medium), NF-M160 kDa neurofilament protein, NFMNeurofilament triplet M protein
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of P07197, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cell Biology, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 4741
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.