missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFkB p105/p50 Antibody (2J10D7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NB100-56583
This item is not returnable.
View return policy
Description
NFkB p105/p50 Monoclonal specifically detects NFkB p105/p50 in Human, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.
Specifications
| NFkB p105/p50 | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 2-5 ug/ml, Flow Cytometry, Immunohistochemistry-Paraffin 5 ug/ml, Knockout Validated | |
| DKFZp686C01211, DNA binding factor KBF1, DNA-binding factor KBF1, EBP-1, KBF1, NF-kappaB, NF-kappa-B, NF-kappabeta, NFKB-p105, NFKB-p50, nuclear factor kappa-B DNA binding subunit, nuclear factor NF-kappa-B p105 subunit, nuclear factor NF-kappa-B p50 subunit, nuclear factor of kappa light polypeptide gene enhancer in B-cells 1MGC54151, p105, p50 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Cross reacts with Human. Predicted to react with Mouse and Rat which are 86% homologous with the immunogen. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoassay | |
| 2J10D7 | |
| Unconjugated | |
| P25942 | |
| NFKB1 | |
| A portion of amino acids 150-200 of human NF-kB (p50) was used as the immunogen. | |
| 0.1 mg | |
| Apoptosis, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
| 4790 | |
| Human, Rat, Rabbit | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction