missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFkB p105/p50 Antibody (2J10D7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
195.00 € - 534.00 €
Specifications
| Antigen | NFkB p105/p50 |
|---|---|
| Clone | 2J10D7 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2-5 ug/ml, Flow Cytometry, Immunohistochemistry-Paraffin 5 ug/ml, Knockout Validated |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoassay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18006295
|
Novus Biologicals
NB100-56583SS |
0.025 mg |
195.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18788453
|
Novus Biologicals
NB100-56583 |
0.1 mg |
534.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NFkB p105/p50 Monoclonal specifically detects NFkB p105/p50 in Human, Rat, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
| NFkB p105/p50 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoassay | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cell Biology, Neurodegeneration, Neuroscience, Signal Transduction, Transcription Factors and Regulators | |
| P25942 | |
| 4790 | |
| A portion of amino acids 150-200 of human NF-kB (p50) was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 2J10D7 | |
| Western Blot 2-5 ug/ml, Flow Cytometry, Immunohistochemistry-Paraffin 5 ug/ml, Knockout Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat, Rabbit | |
| DKFZp686C01211, DNA binding factor KBF1, DNA-binding factor KBF1, EBP-1, KBF1, NF-kappaB, NF-kappa-B, NF-kappabeta, NFKB-p105, NFKB-p50, nuclear factor kappa-B DNA binding subunit, nuclear factor NF-kappa-B p105 subunit, nuclear factor NF-kappa-B p50 subunit, nuclear factor of kappa light polypeptide gene enhancer in B-cells 1MGC54151, p105, p50 | |
| NFKB1 | |
| IgG1 κ | |
| Protein G purified | |
| Cross reacts with Human. Predicted to react with Mouse and Rat which are 86% homologous with the immunogen. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title