missing translation for 'onlineSavingsMsg'
Learn More

Nicergoline, LoGiCal

Product Code. 30461828
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461828 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461828 Supplier LoGiCal Supplier No. LGCFOR0115.00
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nicergoline
CAS 27848-84-6
Analyte or Component Names Nicergoline
Shipping Condition Room Temperature
Molecular Formula C24 H26 Br N3 O3
Certifications/Compliance ISO 9001
Synonym Nicergoline, Ergoline-8-methanol, 10-methoxy-1,6-dimethyl-, 8-(5-bromo-3-pyridinecarboxylate), (8beta)-, Ergoline-8-methanol, 10-methoxy-1,6-dimethyl-, 5-bromo-3-pyridinecarboxylate (ester), (8beta)- (9CI), Ergoline-8beta-methanol, 10-methoxy-1,6-dimethyl-, 5-bromonicotinate (ester) (8CI), Indolo[4,3-fg]quinoline, ergoline-8-methanol deriv., Nicotinic acid, 5-bromo-, (10-methoxy-1,6-dimethylergolin-8beta-yl)methyl ester (8CI), 1,6-Dimethyl-8beta-(5-bromonicotinoyloxymethyl)-10alpha-methoxyergoline, Cergodum, Circo-Maren, Dilasenil, Duracebrol, Ergobel, Ergotop, FI 6714, MNE, Memoq, NSC 150531, Nicergolent, Nicotergoline, Sermion, TA 079, Vasospan
InChI Formula InChI=1S/C24H26BrN3O3/c1-27-13-17-8-21-24(30-3,19-5-4-6-20(27)22(17)19)9-15(12-28(21)2)14-31-23(29)16-7-18(25)11-26-10-16/h4-7,10-11,13,15,21H,8-9,12,14H2,1-3H3/t15-,21-,24+/m1/s1
Solution Type Neat
SMILES CO[C@]12C[C@@H](COC(=O)c3cncc(Br)c3)CN(C)[C@@H]1Cc4cn(C)c5cccc2c45
IUPAC Name [(6aR,9R,10aS)-10a-methoxy-4,7-dimethyl-6a,8,9,10-tetrahydro-6H-indolo[4,3-fg]quinoline-9-yl]methyl 5-bromopyridine-3-carboxylate
Industry Type Clinical Testing
Molecular Weight (g/mol) 484.39
Quantity 10 mg
Formula Weight 483.1158 g/mol
Show More Show Less
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.