missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicergoline, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Nicergoline
Specifications
Specifications
| Chemical Name or Material | Nicergoline |
| CAS | 27848-84-6 |
| Analyte or Component Names | Nicergoline |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H26 Br N3 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Nicergoline, Ergoline-8-methanol, 10-methoxy-1,6-dimethyl-, 8-(5-bromo-3-pyridinecarboxylate), (8beta)-, Ergoline-8-methanol, 10-methoxy-1,6-dimethyl-, 5-bromo-3-pyridinecarboxylate (ester), (8beta)- (9CI), Ergoline-8beta-methanol, 10-methoxy-1,6-dimethyl-, 5-bromonicotinate (ester) (8CI), Indolo[4,3-fg]quinoline, ergoline-8-methanol deriv., Nicotinic acid, 5-bromo-, (10-methoxy-1,6-dimethylergolin-8beta-yl)methyl ester (8CI), 1,6-Dimethyl-8beta-(5-bromonicotinoyloxymethyl)-10alpha-methoxyergoline, Cergodum, Circo-Maren, Dilasenil, Duracebrol, Ergobel, Ergotop, FI 6714, MNE, Memoq, NSC 150531, Nicergolent, Nicotergoline, Sermion, TA 079, Vasospan |
| InChI Formula | InChI=1S/C24H26BrN3O3/c1-27-13-17-8-21-24(30-3,19-5-4-6-20(27)22(17)19)9-15(12-28(21)2)14-31-23(29)16-7-18(25)11-26-10-16/h4-7,10-11,13,15,21H,8-9,12,14H2,1-3H3/t15-,21-,24+/m1/s1 |
| SMILES | CO[C@]12C[C@@H](COC(=O)c3cncc(Br)c3)CN(C)[C@@H]1Cc4cn(C)c5cccc2c45 |
| IUPAC Name | [(6aR,9R,10aS)-10a-methoxy-4,7-dimethyl-6a,8,9,10-tetrahydro-6H-indolo[4,3-fg]quinoline-9-yl]methyl 5-bromopyridine-3-carboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction