missing translation for 'onlineSavingsMsg'
Learn More

Nicotinamide, LoGiCal

Product Code. 30457394
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457394 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457394 Supplier LoGiCal Supplier No. LGCFOR0552.00
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nicotinamide (Vitamin B3)
CAS 98-92-0
Analyte or Component Names Nicotinamide (Vitamin B3)
Shipping Condition Room Temperature
Molecular Formula C6 H6 N2 O
Certifications/Compliance ISO 9001
Synonym Nicotinamide, 3-Pyridinecarboxamide, 3-(Aminocarbonyl)pyridine, 3-Amidopyridine, 3-Carbamoylpyridine, 3-Pyridinecarboxylic acid amide, Aminicotin, Benicot, Delonin Amide, Dipegyl, NAM, NSC 13128, NSC 27452, Niacinamide, Niavit PP, Nicamina, Nicamindon, Nicasir, Nicobion, Nicofort, Nicosan 2, Nicosylamide, Nicotilamide, Nicotine acid amide, Nicotinic acid amide, Nicotinic amide, Nicotylamide, Nicovit, Nicovitina, Nictoamide, Niocinamide, Niozymin, Papulex, Pelmin, Pelmine, Pelonin amide, Vi-Nicotyl, Vitamin B, Vitamin B3, m-(Aminocarbonyl)pyridine, beta-Pyridinecarboxamide
InChI Formula InChI=1S/C6H6N2O/c7-6(9)5-2-1-3-8-4-5/h1-4H,(H2,7,9)
Solution Type Neat
SMILES NC(=O)c1cccnc1
IUPAC Name pyridine-3-carboxamide
Industry Type Clinical Testing
Molecular Weight (g/mol) 122.12
Quantity 10 mg
Formula Weight 122.048 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.