missing translation for 'onlineSavingsMsg'
Learn More

Nicotine [for single use], Mikromol™

Product Code. 17968379
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17968379 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17968379 Supplier Mikromol Supplier No. MM0517.000100
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Nicotine [for single use]
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nicotine
CAS 54-11-5
Analyte or Component Names Nicotine
Shipping Condition Ice pack (-20°C)
Molecular Formula C10 H14 N2
Certifications/Compliance ISO 17034
Synonym Nicotine, (-)-Nicotine, (-)-(S)-Nicotine, (-)-3-(1-Methyl-2-pyrrolidyl)pyridine, (S)-(-)-Nicotine, (S)-3-(1-Methyl-2-pyrrolidinyl)pyridine, (S)-Nicotine, 3-[(2S)-1-Methyl-2-pyrrolidinyl]pyridine, Exodus, Flux Maag, Habitrol, L-Nicotine, NSC 5065, Nicabate, Nicoderm, Nicolan, Niconil, Nicopatch, Nicorette, Nicotell TTS, Nicotin, Nicotinell, Nicotrol, Nikofrenon, Niquitin, Tabazur, XL All Insecticide, l-Nicotine, Nicotine (8CI), Pyridine, 3-(1-methyl-2-pyrrolidinyl)-, (S)-, (-)-beta-Pyridyl-alpha-N-methylpyrrolidine
InChI Formula InChI=1S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10H,3,5,7H2,1H3/t10-/m0/s1
SMILES CN1CCC[C@H]1c2cccnc2
IUPAC Name 3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
Molecular Weight (g/mol) 162.23
Quantity 100 mg
Formula Weight 162.1157 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.