missing translation for 'onlineSavingsMsg'
Learn More

Nicotinic Acid 1.0 mg/ml in Methanol, LoGiCal

Product Code. 30458801
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458801 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458801 Supplier LoGiCal Supplier No. LGCAMP0553.0001
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Niacin (Nicotinic Acid)
CAS 59-67-6
Analyte or Component Names Niacin (Nicotinic Acid)
Shipping Condition Ice pack (-20°C)
Molecular Formula C6 H5 N O2
Certifications/Compliance ISO 17034
Synonym Nicotinic Acid, Methyl Nicotinate Imp. A (EP), 3-Pyridinecarboxylic acid, Nicotinic acid, 3-Carboxylpyridine, 3-Carboxypyridine, 3-Pyridylcarboxylic acid, Akotin, Apelagrin, CaRo 03, Daskil, E 375, Efacin, Enduracin, Linic, Microvit B3 Promix Niacin, NSC 169454, Niac, Niacin, Niacor, Niaspan, Nicacid, Nicangin, Nico-Span, Nicobid, Nicodelmine, Nicolar, Niconacid, Nicosan 3, Nicotinipca, Nicyl, Nyclin, Pellagrin, Pelonin, SR 4390, Slo-niacin, Vitamin B3, Vitamin B5, Wampocap, beta-Pyridinecarboxylic acid, Methyl Nicotinate Impurity A
InChI Formula InChI=1S/C6H5NO2/c8-6(9)5-2-1-3-7-4-5/h1-4H,(H,8,9)
Solution Type Single Solution
SMILES OC(=O)c1cccnc1
IUPAC Name pyridine-3-carboxylic acid
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 123.11
Quantity 1 mL
Formula Weight 123.032 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.