missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Niemann-Pick C1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97133-50ul
This item is not returnable.
View return policy
Description
Niemann-Pick C1 Polyclonal antibody specifically detects Niemann-Pick C1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| Niemann-Pick C1 | |
| Polyclonal | |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:200-1:500 | |
| FLJ98532, Niemann-Pick C1 protein, Niemann-Pick disease, type C1, NPC, NPC1 | |
| Produced in rabbits immunized with purified, recombinant Human Niemann-Pick C1 (Accession#: NP_000262.2; Arg372-Phe622) | |
| 50 μg | |
| Cancer, Cholesterol Metabolism, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction | |
| 4864 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur