missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Niemann-Pick C1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 339.00 €
Specifications
| Antigen | Niemann-Pick C1 |
|---|---|
| Dilution | ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:200-1:500 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18644732
|
Novus Biologicals
NBP2-97133-100ul |
100 μg |
339.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18675332
|
Novus Biologicals
NBP2-97133-50ul |
50 μg |
216.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Niemann-Pick C1 Polyclonal antibody specifically detects Niemann-Pick C1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
| Niemann-Pick C1 | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cholesterol Metabolism, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 4864 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:200-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| FLJ98532, Niemann-Pick C1 protein, Niemann-Pick disease, type C1, NPC, NPC1 | |
| Produced in rabbits immunized with purified, recombinant Human Niemann-Pick C1 (Accession#: NP_000262.2; Arg372-Phe622) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title