missing translation for 'onlineSavingsMsg'
Learn More

Nifedipine, Mikromol™

Product Code. 17962226
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17962226 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17962226 Supplier Mikromol Supplier No. MM0003.000250
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Nifedipine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nifedipine
CAS 21829-25-4
Analyte or Component Names Nifedipine
Shipping Condition Room Temperature
Molecular Formula C17 H18 N2 O6
Certifications/Compliance ISO 17034
Synonym 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(2-nitrophenyl)-, 3,5-dimethyl ester, 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(2-nitrophenyl)-, dimethyl ester, 3,5-Pyridinedicarboxylic acid, 1,4-dihydro-2,6-dimethyl-4-(o-nitrophenyl)-, dimethyl ester, 2,6-Dimethyl-3,5-dicarbomethoxy-4-(2-nitrophenyl)-1,4-dihydropyridine, 2,6-Dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylic acid dimethyl ester, 4-(2-Nitrophenyl)-2,6-dimethyl-3,5-dicarbomethoxy-1,4-dihydropyridine, Adalat, Adalat 10, Adalat 20, Adalat 5, Adalat CC, Adalat CR, Adalat Crono, Adalat FT, Adalat GITS, Adalat GITS 30, Adalat LA, Adalat LP, Adalat Oros, Adalat PA, Adalat retard, Adalate, Adapine, Adapress, Alat, Aldipin, Alfadat, Alonix, Alonix S, Alpha-Nifedipine Retard, Angipec, Anifed, Anpine, Apo-Nifed, Aprical, BAY 1040, BAY-a 1040, Bonacid, Calcibloc, Calcigard, Calcilat, Camont, Cardifen, Cardilat, Cardilate, Cardionorm, Chronadalate, Chronadalate LP, Citilat, Coracten, Coral, Cordafen, Cordaflex, Cordalat, Cordicant, Cordilan, Cordipin, Corinfar, Corotrend, Corynphar, Depin, Dignokonstant, Dilafed, Dilcor, Dimethyl 1,4-dihydro-2,6-dimethyl-4-(2-nitrophenyl)-3,5-pyridinedicarboxylate, Dimethyl 4-(o-Nitrophenyl)-2,6-dimethyl-1,4-dihydro-3,5-pyridinedicarboxylate, Dipinkor, Duranifin, Ecodipi, Ecodipin, Ecodipin E, Fedcor, Fedcor Retard, Fedipin, Fenamon, Fenamon SR, Fenihidine, Glopir, Hadipine, Hexadilat, Introcar, Kordafen, Macorel, Megalat, Myogard, NIfedilat, Nedipin, Nicardia, Nifangin, Nifar, Nifdemin, Nifebene, Nifecard, Nifecor, Nifedate, Nifedepat, Nifedicor, Nifedin, Nifedine, Nifedipine, Nifedipine retard, Nifedipres, Nifedirex LP, Nifelan, Nifelat, Nifelat Q, Nifelate, Nifensar XL, Nificard, Nifidine, Nifipen, Niphedipine, Normadil, Novo Nifedin, Nyefax, Nyefax Retard, Orix, Osmo-Adalat, Oxcord, Pidilat, Procardia, Procardia XL, Sepamit, Sepamit R, Slofedipine XL, TN 873R, Tensopin, Tibricol, UV-ENP 349PINA, Vasdalat, Vasdalat Retard, Vasofed, Zenusin
InChI Formula InChI=1S/C17H18N2O6/c1-9-13(16(20)24-3)15(14(10(2)18-9)17(21)25-4)11-7-5-6-8-12(11)19(22)23/h5-8,15,18H,1-4H3
SMILES COC(=O)C1=C(C)NC(=C(C1c2ccccc2[N+](=O)[O-])C(=O)OC)C
IUPAC Name dimethyl 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Weight (g/mol) 346.33
Quantity 250 mg
Formula Weight 346.1165 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.