missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NIPA Polyclonal antibody specifically detects NIPA in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | NIPA |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | C3HC type 1, FBXO50, zinc finger, C3HC-type containing 1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human ZC3HC1 (NP_057562.3). MAAPCEGQAFAVGVEKNWGAVVRSPEGTPQKIRQLIDEGIAPEEGGVDAKDTSATSQSVNGSPQAEQPSLESTSKEAFFSRVETFSSLKWAGKPFELSPLVCAKYGWVTVECDMLKCSSCQAFLCASLQPAFDFDRYKQRCAELKKALCTAHEKFCFWPDSPSPDRFGMLPLDEPAILVSEFLDRFQSLCHLDLQLPSLRPEDLKTMCLTEDKISLLLHLLEDELDHRTDERKTTIKLGSDIQVHVTACILSVCGWACSS |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?