missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRC3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41123-0.025mg
This item is not returnable.
View return policy
Description
NLRC3 Polyclonal specifically detects NLRC3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence.
Specifications
| NLRC3 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| NLRC3 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the center of human NOD3. The immunogen is located within amino acids 630 - 680 of NOD3. Amino Acid Squence: CRKLRLDTNQFQDPVMEL | |
| 0.025 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1 - 2 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 20 μg/mL | |
| CARD15-Like Protein, Caterpiller Protein 16.2, CLR16.2, Leucine Rich Repeat And CARD Domain Containing 3, NLR Family, CARD Domain Containing 3, NOD3, NOD-Like Receptor C3, Nucleotide-Binding Oligomerization Domain Protein 3, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And CARD Domain Containing 3, Protein NLRC3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 197358 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction