missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRC3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
189.00 € - 523.00 €
Specifications
| Antigen | NLRC3 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
|
18636025
|
Novus Biologicals
NBP2-41123-0.025mg |
0.025 mg |
189.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18010434
|
Novus Biologicals
NBP2-41123 |
0.1 mg |
523.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
NLRC3 Polyclonal specifically detects NLRC3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence.Especificaciones
| NLRC3 | |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 197358 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the center of human NOD3. The immunogen is located within amino acids 630 - 680 of NOD3. Amino Acid Squence: CRKLRLDTNQFQDPVMEL | |
| Primary |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| CARD15-Like Protein, Caterpiller Protein 16.2, CLR16.2, Leucine Rich Repeat And CARD Domain Containing 3, NLR Family, CARD Domain Containing 3, NOD3, NOD-Like Receptor C3, Nucleotide-Binding Oligomerization Domain Protein 3, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And CARD Domain Containing 3, Protein NLRC3 | |
| NLRC3 | |
| IgG | |
| Affinity Purified |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto