missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-27198SS
This item is not returnable.
View return policy
Description
NLRX1 Polyclonal specifically detects NLRX1 in Human, Mouse, Rat, Equine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| NLRX1 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Caterpiller protein 11.3, CLR11.3DLNB26, MGC131937, MGC21025, NLR family member X1, NLR family, X1, NOD26, NOD5, NOD9FLJ21478, NOD-like receptor X1, Nucleotide-binding oligomerization domain protein 26, Nucleotide-binding oligomerization domain protein 5, Nucleotide-binding oligomerization domain protein 9, nucleotide-binding oligomerization domain, leucine rich repeat containing X1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 79671 | |
| Human, Mouse, Rat, Equine, Monkey | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1.0 mg/ml | |
| Western Blot 5-7ug/ml∼, Immunohistochemistry-Paraffin 5-10ug/ml | |
| Q86UT6 | |
| NLRX1 | |
| A portion of amino acid 580-630 of human NLRX1 was used as the immunogen for the antibody. | |
| 0.025 mg | |
| Primary | |
| The amino acid sequence used as immunogen for the NLRX1 antibody is 100% homologous in human, mouse, rat, monkey and horse, and 95% homologous in cow and dog. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction