missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 497.00 €
Specifications
| Antigen | NLRX1 |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 5-7ug/ml∼, Immunohistochemistry-Paraffin 5-10ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18138363
|
Novus Biologicals
NBP2-27198SS |
0.025 mg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18725853
|
Novus Biologicals
NBP2-27198 |
0.1 mg |
497.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NLRX1 Polyclonal specifically detects NLRX1 in Human, Mouse, Rat, Equine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NLRX1 | |
| Western Blot 5-7ug/ml∼, Immunohistochemistry-Paraffin 5-10ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Caterpiller protein 11.3, CLR11.3DLNB26, MGC131937, MGC21025, NLR family member X1, NLR family, X1, NOD26, NOD5, NOD9FLJ21478, NOD-like receptor X1, Nucleotide-binding oligomerization domain protein 26, Nucleotide-binding oligomerization domain protein 5, Nucleotide-binding oligomerization domain protein 9, nucleotide-binding oligomerization domain, leucine rich repeat containing X1 | |
| NLRX1 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen for the NLRX1 antibody is 100% homologous in human, mouse, rat, monkey and horse, and 95% homologous in cow and dog. |
| 1.0 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat, Equine, Monkey | |
| Q86UT6 | |
| 79671 | |
| A portion of amino acid 580-630 of human NLRX1 was used as the immunogen for the antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title