missing translation for 'onlineSavingsMsg'
Learn More

NME5 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18533354 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18533354 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18533354 Supplier Novus Biologicals Supplier No. NBP298490AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NME5 Polyclonal antibody specifically detects NME5 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen NME5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias Inhibitor of p53-induced apoptosis-beta, IPIA-beta, NDK-H 5, NDP kinase homolog 5, nm23-H5, NM23H5, non-metastatic cells 5, protein expressed in (nucleoside-diphosphate kinase), nucleoside diphosphate kinase homolog 5, radial spoke 23 homolog, RSPH23, Testis-specific nm23 homolog
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human NME5 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8382
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.