missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB100-56878SS
This item is not returnable.
View return policy
Description
NOD1 Polyclonal specifically detects NOD1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| NOD1 | |
| Polyclonal | |
| Western Blot 1:1000-1:2500, Immunohistochemistry-Paraffin 1:100 | |
| Q9Y239 | |
| NOD1 | |
| Amino acids 930-949 of human NOD1 protein were used as the immunogen for this antibody. | |
| 0.025 mL | |
| Apoptosis, Asthma, Autophagy, Immunology | |
| 10392 | |
| Human | |
| Antisera |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with 50% Glycerol with 0.025% Sodium Azide | |
| CARD4NLR family, CARD domain containing 1, caspase recruitment domain family, member 4, Caspase recruitment domain-containing protein 4, CLR7.1, NLRC1, nucleotide-binding oligomerization domain containing 1, nucleotide-binding oligomerization domain, leucine rich repeat and CARD domaincontaining 1, nucleotide-binding oligomerization domain-containing protein 1 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Cross reacts with Human. Predicted to cross react with New World Monkey and Mouse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction