missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
213.00 € - 390.00 €
Specifications
| Antigen | NOD1 |
|---|---|
| Dilution | Western Blot 1:1000-1:2500, Immunohistochemistry-Paraffin 1:100 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18031264
|
Novus Biologicals
NB100-56878SS |
0.025 mL |
213.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18747653
|
Novus Biologicals
NB100-56878 |
390.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
NOD1 Polyclonal specifically detects NOD1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NOD1 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Asthma, Autophagy, Immunology | |
| Whole antisera with 50% Glycerol with 0.025% Sodium Azide | |
| CARD4NLR family, CARD domain containing 1, caspase recruitment domain family, member 4, Caspase recruitment domain-containing protein 4, CLR7.1, NLRC1, nucleotide-binding oligomerization domain containing 1, nucleotide-binding oligomerization domain, leucine rich repeat and CARD domaincontaining 1, nucleotide-binding oligomerization domain-containing protein 1 | |
| NOD1 | |
| IgG | |
| Unpurified | |
| Cross reacts with Human. Predicted to cross react with New World Monkey and Mouse. |
| Western Blot 1:1000-1:2500, Immunohistochemistry-Paraffin 1:100 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human | |
| Q9Y239 | |
| 10392 | |
| Amino acids 930-949 of human NOD1 protein were used as the immunogen for this antibody. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title