missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nogo-A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41071-0.025mg
This item is not returnable.
View return policy
Description
Nogo-A Polyclonal specifically detects Nogo-A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Nogo-A | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Rabbit | |
| ∼180 kDa | |
| 0.025 mg | |
| Primary | |
| At least five isoforms of Nogo are known to exist; this antibody is specific for NogoA and NogoE. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 0.5 - 1 μg/mL, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| RTN4 | |
| Antibody was raised against a 19 amino acid synthetic peptide from near the center of human NogoA. The immunogen is located within amino acids 200 - 250 of NogoA. Amino Acid Squence: DLKEQPGNTISAGQEDFPS | |
| Affinity Purified | |
| RUO | |
| 57142 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction